2-(Pyrimidin-5-yl)ethanol 97%

Supplier: Ambeed
Warning

A286292-1G A286292-250MG A286292-100MG
76787-464EA 176 USD
76787-464 76787-466 77872-363
2-(Pyrimidin-5-yl)ethanol 97%
2-(Pyrimidin-5-yl)ethanol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD09926205
CAS Number: 875251-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR