trans-2-Hexenoic acid 97%

Supplier: Ambeed
Danger

Synonyms: trans-Hex-2-enoic acid

A284709-1000G A284709-100G A284709-10G A284709-25G A284709-500G
76787-446EA 267.35 USD
76787-446 76787-448 76787-450 76787-452 76787-454
trans-2-Hexenoic acid 97%
trans-2-Hexenoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00002705
CAS Number: 13419-69-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR