Ethyl-3-(4-bromophenyl)propanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(4-bromophenyl)propanoate, Ethyl 3-(4-bromophenyl)propionate

A283657-10G A283657-1G A283657-25G A283657-5G A283657-100G
76787-430EA 29 USD
76787-430 76787-432 76787-434 76787-436 77872-360
Ethyl-3-(4-bromophenyl)propanoate 98%
Ethyl-3-(4-bromophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MDL Number: MFCD07772961
CAS Number: 40640-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR