trans-2,5-Dioxopyrrolidin-1-yl 4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Trans-4-(Maleimidomethyl)cyclohexanecarboxylic Acid-NHS

A283587-100MG A283587-10MG A283587-1MG A283587-50MG A283587-250MG
76787-422EA 32.89 USD
76787-422 76787-424 76787-426 76787-428 77668-252
trans-2,5-Dioxopyrrolidin-1-yl 4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carboxylate 95%
trans-2,5-Dioxopyrrolidin-1-yl 4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₂O₆
MW: 334.32 g/mol
MDL Number: MFCD08274640
CAS Number: 71875-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR