trans-2-Hexen-1-ol 95%

Supplier: Ambeed
Danger

A282580-25G A282580-5G
76787-418EA 8 USD
76787-418 76787-420
trans-2-Hexen-1-ol 95%
trans-2-Hexen-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₂O
MDL Number: MFCD00002927
CAS Number: 928-95-0
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR