Terazosin hydrochloride dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6,7-dimethoxy-2-[4-(tetrahydro-2-furoyl)-1-piperazinyl]quinazoline Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(tetrahydrofuran-2-yl)methanone hydrochloride dihydrate

A281463-100MG A281463-10G A281463-1G
76787-408EA 5.31 USD
76787-408 76787-410 76787-412
Terazosin hydrochloride dihydrate 98%
Terazosin hydrochloride dihydrate

Precaución: for Research Use Only

Formula: C₁₉H₃₀ClN₅O₆
MW: 459.92 g/mol
MDL Number: MFCD00941408
CAS Number: 70024-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR