Penta-2,4-dienoic acid ≥95% (by GC, sum of isomers)

Supplier: Ambeed
Danger

A281102-1G A281102-250MG A281102-5G
76787-390EA 146 USD
76787-390 76787-392 76787-394
Penta-2,4-dienoic acid ≥95% (by GC, sum of isomers)
Penta-2,4-dienoic acid

Precaución: for Research Use Only

Formula: C₅H₆O₂
MDL Number: MFCD00014017
CAS Number: 626-99-3
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR