N-Benzyloxycarbonyl-L-valinamide 97%

Supplier: Ambeed
Warning

Synonyms: Z-Val-NH₂, Z-Val-NH2, (S)-Benzyl (1-amino-3-methyl-1-oxobutan-2-yl)carbamate

A280312-1G A280312-250MG A280312-25G
76787-384EA 23 USD
76787-384 76787-386 76787-388
N-Benzyloxycarbonyl-L-valinamide 97%
N-Benzyloxycarbonyl-L-valinamide

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₃
MW: 250.298 g/mol
MDL Number: MFCD00191170
CAS Number: 13139-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR