4'-Methyl-[1,1'-biphenyl]-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A279918-1G A279918-5G
76787-376EA 103.97 USD
76787-376 76787-378
4'-Methyl-[1,1'-biphenyl]-3-carbaldehyde 97%
4'-Methyl-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.24 g/mol
MDL Number: MFCD03424625
CAS Number: 116470-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR