Fmoc-D-allo-thr(tBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-O-tert-butyl-D-allo-threonine, Fmoc-D-Allo-Thr(tBu)-OH

A278682-100MG A278682-250MG A278682-1G A278682-5G
76787-360EA 21 USD
76787-360 76787-362 77571-732 77571-734
Fmoc-D-allo-thr(tBu)-OH 98%
Fmoc-D-allo-thr(tBu)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₅
MW: 397.471 g/mol
MDL Number: MFCD00077074
CAS Number: 170643-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR