Z-Ala-OSu 95%

Supplier: Ambeed
Warning

A278382-100G A278382-1G A278382-5G A278382-25G A278382-500G
76787-354EA 180 USD
76787-354 76787-356 76787-358 77668-216 77668-218
Z-Ala-OSu 95%
Z-Ala-OSu

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O₆
MDL Number: MFCD00053548
CAS Number: 3401-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR