8-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2,4-dimethyl-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-one 95%

Supplier: Ambeed
Danger

A278302-10MG A278302-1MG A278302-5MG
76787-348EA 169.75 USD
76787-348 76787-350 76787-352
8-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2,4-dimethyl-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-one 95%
8-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2,4-dimethyl-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-one

Precaución: for Research Use Only

Formula: C₂₃H₂₁N₇O
MW: 411.45 g/mol
MDL Number: MFCD00865905
CAS Number: 145733-36-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR