Manganese(II) acetate tetrahydrate 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Manganese acetate tetrahydrate, Manganese diacetate tetrahydrate, Acetic acid manganese (II) salt tetrahydrate
A278087-1000G
A278087-100G
A278087-10G
A278087-25G
A278087-500G
76787-338EA
19
USD
76787-338
76787-340
76787-342
76787-344
76787-346
Manganese(II) acetate tetrahydrate 99+%
Manganese(II) acetate tetrahydrate
Precaución: for Research Use Only
Formula:
(H₃CCOO)₂Mn·4H₂O MW: 245.09 g/mol Melting Pt: 300 |
MDL Number:
MFCD00062552 CAS Number: 6156-78-1 Merck Index: 13,05746 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...