SB202190 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(4-Fluorophenyl)-5-(pyridin-4-yl)-1H-imidazol-2-yl)phenol, SB 202190

A277214-10MG A277214-1MG A277214-25MG A277214-50MG A277214-5MG A277214-100MG A277214-250MG
76787-328EA 64 USD
76787-328 76787-330 76787-332 76787-334 76787-336 77601-224 77872-336
SB202190 98%
SB202190

Precaución: for Research Use Only

Formula: C₂₀H₁₄FN₃O
MW: 331.3 g/mol
MDL Number: MFCD00941964
CAS Number: 152121-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR