3-(3-(Trifluoromethyl)phenoxy)benzaldehyde 95+%

Supplier: Ambeed
Warning

A277122-1G A277122-250MG A277122-5G
76787-316EA 27.59 USD
76787-316 76787-318 76787-320
3-(3-(Trifluoromethyl)phenoxy)benzaldehyde 95+%
3-(3-(Trifluoromethyl)phenoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
MDL Number: MFCD00003356
CAS Number: 78725-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR