2-Amino-5-bromo-3-ethynylpyrazine 98%

Supplier: Ambeed
Warning

A273470-1G A273470-250MG A273470-5G
76787-224EA 64 USD
76787-224 76787-226 76787-228
2-Amino-5-bromo-3-ethynylpyrazine 98%
2-Amino-5-bromo-3-ethynylpyrazine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MDL Number: MFCD09999142
CAS Number: 1209289-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR