3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Warning

A272471-250MG A272471-1G
76787-220EA 76.39 USD
76787-220 77601-168
3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine 97%
3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
MDL Number: MFCD11007847
CAS Number: 947238-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR