Zinc(II) acetylacetonate 98%

Supplier: Ambeed
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II), Zinc acetylacetonate, Zinc (II) 2,4-pentanedionate, Zinc (II) bis(2,4-pentanedionato-O,O')

A272165-1000G A272165-100G A272165-25G A272165-500G
76787-212EA 43.5 USD
76787-212 76787-214 76787-216 76787-218
Zinc(II) acetylacetonate 98%
Zinc(II) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄Zn
MW: 263.61 g/mol
MDL Number: MFCD00000035
CAS Number: 14024-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR