4'-Bromo-[1,1'-biphenyl]-4-carbonitrile 95%

Supplier: Ambeed
Warning

A270749-1G A270749-25G
76787-180EA 13 USD
76787-180 77668-182
4'-Bromo-[1,1'-biphenyl]-4-carbonitrile 95%
4'-Bromo-[1,1'-biphenyl]-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈BrN
MDL Number: MFCD05980943
CAS Number: 57774-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR