1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol 97%

Supplier: Ambeed
Warning

A269929-1G A269929-250MG A269929-5G
76787-166EA 157.02 USD
76787-166 76787-168 76787-170
1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol 97%
1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol

Precaución: for Research Use Only

Formula: C₁₈H₂₂O₆
MW: 334.3600 g/mol
MDL Number: MFCD01880862
CAS Number: 10535-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR