2-Acetylphenothiazine 98%

Supplier: Ambeed
Warning

A269257-100G A269257-100MG A269257-1G A269257-25MG A269257-5G A269257-25G A269257-1MG
76787-150EA 487 USD
76787-150 76787-152 76787-154 76787-156 76787-158 77601-152 77668-174
2-Acetylphenothiazine 98%
2-Acetylphenothiazine

Precaución: for Research Use Only

Formula: C₁₄H₁₁NOS
MDL Number: MFCD00005017
CAS Number: 6631-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR