Methyl-1-boc-3-azetidineacetate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-methoxy-2-oxoethyl)azetidine-1-carboxylate

A269179-10G A269179-1G A269179-25G A269179-5G
76787-140EA 100 USD
76787-140 76787-142 76787-144 76787-146
Methyl-1-boc-3-azetidineacetate 97%
Methyl-1-boc-3-azetidineacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD11559001
CAS Number: 497160-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR