(1-Methyl-1H-benzo[d]imidazol-5-yl)methanol 96%

Supplier: Ambeed
Warning

A268745-1G A268745-250MG A268745-5G
76787-136EA 81 USD
76787-136 76787-138 77872-310
(1-Methyl-1H-benzo[d]imidazol-5-yl)methanol 96%
(1-Methyl-1H-benzo[d]imidazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
MDL Number: MFCD11616105
CAS Number: 115576-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR