Theacrine 97%

Supplier: Ambeed
Warning

Synonyms: Temorine, 1,3,7,9-Tetramethyluric acid, 1,3,7,9-Tetramethyl-1H-purine-2,6,8(3H,7H,9H)-trione

A266973-1G A266973-250MG A266973-25G A266973-5G
76787-104EA 9.55 USD
76787-104 76787-106 76787-108 76787-110
Theacrine 97%
Theacrine

Precaución: for Research Use Only

Formula: C₉H₁₂N₄O₃
MW: 224.2 g/mol
MDL Number: MFCD00042781
CAS Number: 2309-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR