4-Chloro-5-fluoro-2-pyridinemethanol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-5-fluoropyridin-2-yl)methanol

A266710-100MG A266710-250MG A266710-5G
76787-092EA 109 USD
76787-092 76787-094 76787-096
4-Chloro-5-fluoro-2-pyridinemethanol 97%
4-Chloro-5-fluoro-2-pyridinemethanol

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MW: 161.56 g/mol
MDL Number: MFCD09953777
CAS Number: 113209-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR