5-Methoxypyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxypicolinonitrile, 2-Cyano-5-methoxypyridine

A265301-1G A265301-250MG A265301-5G A265301-100MG
76787-072EA 106.09 USD
76787-072 76787-074 76787-076 77668-140
5-Methoxypyridine-2-carbonitrile 98%
5-Methoxypyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.135 g/mol
MDL Number: MFCD08543467
CAS Number: 89809-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR