2-(3-Bromophenyl)-1H-imidazole 98%

Supplier: Ambeed
Warning

A265088-100MG A265088-250MG
76787-062EA 42.44 USD
76787-062 76787-064
2-(3-Bromophenyl)-1H-imidazole 98%
2-(3-Bromophenyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.06 g/mol
MDL Number: MFCD08668744
CAS Number: 937013-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR