(1S,4S)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride 97%

Supplier: Ambeed
Warning

A265044-10G A265044-1G A265044-250MG A265044-25G A265044-5G
76787-052EA 177.18 USD
76787-052 76787-054 76787-056 76787-058 76787-060
(1S,4S)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride 97%
(1S,4S)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO
MW: 135.5920 g/mol
MDL Number: MFCD01569249
CAS Number: 31560-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR