4,6-Dichloro-2-(trifluoromethyl)pyrimidine 98%

Supplier: Ambeed
Danger

A263679-100MG A263679-10G A263679-250MG A263679-5G A263679-1G
76787-040EA 11.67 USD
76787-040 76787-042 76787-044 76787-046 77571-634
4,6-Dichloro-2-(trifluoromethyl)pyrimidine 98%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅HCl₂F₃N₂
MW: 216.98 g/mol
MDL Number: MFCD08436597
CAS Number: 705-24-8
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR