Tetraethylene glycol monooctyl ether 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
n-Octyltetraoxyethylene, 3,6,9,12-Tetraoxaicosan-1-ol, 3,6,9,12-Tetraoxaeicosan-1-ol
A263432-250MG
A263432-100MG
76787-038EA
7
USD
76787-038
77668-132
Tetraethylene glycol monooctyl ether 95+%
Tetraethylene glycol monooctyl ether
Precaución: for Research Use Only
Formula:
C₁₆H₃₄O₅ |
MDL Number:
MFCD00043033 CAS Number: 19327-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...