Ethyl-2,4-dichloronicotinate 98%

Supplier: Ambeed
Warning

A263087-10G A263087-5G A263087-1G A263087-250MG
76787-032EA 189 USD
76787-032 77601-126 77872-290 77872-291
Ethyl-2,4-dichloronicotinate 98%
Ethyl-2,4-dichloronicotinate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MDL Number: MFCD12407388
CAS Number: 62022-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR