trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trifluoro-4'-((trans,trans)-4'-propyl-[1,1'-bi(cyclohexan)]-4-yl)-1,1'-biphenyl

A262834-100G A262834-1G A262834-25G A262834-5G
76787-024EA 389.36 USD
76787-024 76787-026 76787-028 76787-030
trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl 97%
trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl

Precaución: for Research Use Only

Formula: C₂₇H₃₃F₃
MW: 414.56 g/mol
MDL Number: MFCD13188632
CAS Number: 137529-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR