6-Amino-7-bromobenzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 7-bromo-1,3-benzothiazol-6-amine

A261485-100MG A261485-1G A261485-250MG A261485-5G
76786-998EA 22.28 USD
76786-998 76787-000 76787-002 76787-004
6-Amino-7-bromobenzothiazole 98%
6-Amino-7-bromobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂S
MW: 229.1000 g/mol
MDL Number: MFCD03426350
CAS Number: 769-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR