4,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A260048-10G A260048-1G A260048-250MG A260048-25G A260048-5G
76786-974EA 87 USD
76786-974 76786-976 76786-978 76786-980 76786-982
4,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 98%
4,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
MDL Number: MFCD09999173
CAS Number: 115093-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR