2-(((Benzyloxy)carbonyl)amino)-5-((tert-butoxycarbonyl)amino)pentanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Z-DL-Orn(Boc)-OH

A259967-10G A259967-1G A259967-5G
76786-968EA 141.1 USD
76786-968 76786-970 76786-972
2-(((Benzyloxy)carbonyl)amino)-5-((tert-butoxycarbonyl)amino)pentanoic acid 97%
2-(((Benzyloxy)carbonyl)amino)-5-((tert-butoxycarbonyl)amino)pentanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₆
MW: 366.40 g/mol
MDL Number: MFCD00038268
CAS Number: 199924-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR