Tetramethyl orthocarbonate 97%

Supplier: Ambeed
Danger

Synonyms: Tetramethylorthocarbonate

A259874-1G A259874-25G A259874-5G
76786-964EA 22.28 USD
76786-964 76786-966 77668-118
Tetramethyl orthocarbonate 97%
Tetramethyl orthocarbonate

Precaución: for Research Use Only

Formula: C₅H₁₂O₄
MW: 136.15 g/mol
MDL Number: MFCD00008473
CAS Number: 1850-14-2
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR