4-Bromo-2-fluoro-6-nitroaniline 98%

Supplier: Ambeed
Warning

A259516-100G A259516-10G A259516-1G A259516-250MG A259516-5G A259516-25G
76786-948EA 155 USD
76786-948 76786-950 76786-952 76786-954 76786-956 77601-096
4-Bromo-2-fluoro-6-nitroaniline 98%
4-Bromo-2-fluoro-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFN₂O₂
MDL Number: MFCD03094271
CAS Number: 517920-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR