4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

A259124-10G A259124-1G
76786-940EA 297.06 USD
76786-940 76786-942
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 98%
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
MDL Number: MFCD11617811
CAS Number: 32980-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR