Tris(pentafluorophenyl)phosphine 98%

Supplier: Ambeed
Warning

A258548-1G A258548-25G A258548-5G A258548-100G
76786-922EA 11 USD
76786-922 76786-924 76786-926 77872-275
Tris(pentafluorophenyl)phosphine 98%
Tris(pentafluorophenyl)phosphine

Precaución: for Research Use Only

Formula: C₁₈F₁₅P
MDL Number: MFCD00079654
CAS Number: 1259-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR