4'-Trifluoromethylbiphenyl-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxaldehyde, 4-(4-Trifluoromethylphenyl)benzaldehyde, 4'-Trifluoromethyl-biphenyl-4-carbaldehyde, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

A257540-25G A257540-5G A257540-1G
76786-906EA 328.88 USD
76786-906 76786-908 77601-070
4'-Trifluoromethylbiphenyl-4-carbaldehyde 97%
4'-Trifluoromethylbiphenyl-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O
MW: 250.2200 g/mol
MDL Number: MFCD01862519
CAS Number: 90035-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR