1-(2,4-Difluorophenyl)-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Temafloxacin HCl

A257138-100MG A257138-1G A257138-250MG
76786-892EA 343.74 USD
76786-892 76786-894 76786-896
1-(2,4-Difluorophenyl)-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride 97%
1-(2,4-Difluorophenyl)-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₁₉ClF₃N₃O₃
MW: 453.84 g/mol
MDL Number: MFCD00903771
CAS Number: 105784-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR