6-Bromothiazolo[4,5-b]pyrazin-2-amine 97%

Supplier: Ambeed
Warning

A253808-100MG A253808-1G A253808-250MG A253808-5G
76786-854EA 59.42 USD
76786-854 76786-856 76786-858 76786-860
6-Bromothiazolo[4,5-b]pyrazin-2-amine 97%
6-Bromothiazolo[4,5-b]pyrazin-2-amine

Precaución: for Research Use Only

Formula: C₅H₃BrN₄S
MW: 231.0731 g/mol
MDL Number: MFCD09909710
CAS Number: 112342-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR