4,5-Difluoro-2-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A253353-1G A253353-250MG
76786-850EA 22 USD
76786-850 76786-852
4,5-Difluoro-2-methylbenzonitrile 98%
4,5-Difluoro-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MDL Number: MFCD09996935
CAS Number: 1003708-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR