Tetrabromophthalic anhydride 98%

Supplier: Ambeed
Warning

A253030-1000G A253030-100G A253030-25G A253030-500G
76786-838EA 80 USD
76786-838 76786-840 76786-842 76786-844
Tetrabromophthalic anhydride 98%
Tetrabromophthalic anhydride

Precaución: for Research Use Only

Formula: C₈Br₄O₃
MDL Number: MFCD00005919
CAS Number: 632-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR