Sulindac 98%

Supplier: Ambeed
Danger

Synonyms: (Z)-2-(5-Fluoro-2-methyl-1-(4-(methylsulphinyl)benzylidene)-1H-inden-3-yl)acetic acid

A253022-100MG A253022-5G A253022-1G A253022-25G
76786-836EA 5.31 USD
76786-836 77568-448 77601-036 77668-062
Sulindac 98%
Sulindac

Precaución: For research use only.

Formula: C₂₀H₁₇FO₃S
MW: 356.411 g/mol
MDL Number: MFCD00599589
CAS Number: 38194-50-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR