(3aR,4aS,8aR,9aR)-8a-Methyl-3,5-dimethylenedecahydronaphtho[2,3-b]furan-2(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: Isoalantolactone

A249503-10MG A249503-1MG A249503-50MG A249503-5MG A249503-100MG A249503-1G A249503-250MG A249503-5G
76786-788EA 9.55 USD
76786-788 76786-790 76786-792 76786-794 77601-008 77601-010 77668-036 77668-038
(3aR,4aS,8aR,9aR)-8a-Methyl-3,5-dimethylenedecahydronaphtho[2,3-b]furan-2(3H)-one 98%
(3aR,4aS,8aR,9aR)-8a-Methyl-3,5-dimethylenedecahydronaphtho[2,3-b]furan-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₂
MW: 232.32 g/mol
MDL Number: MFCD08689940
CAS Number: 470-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR