4,6-Dichloroimidazo[4,5-c]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloro-1H-imidazo[4,5-c]pyridine

A249380-10G A249380-1G A249380-250MG A249380-25G A249380-5G A249380-100MG
76786-778EA 650.34 USD
76786-778 76786-780 76786-782 76786-784 76786-786 77593-574
4,6-Dichloroimidazo[4,5-c]pyridine 97%
4,6-Dichloroimidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
MDL Number: MFCD09753581
CAS Number: 2589-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR