Tetrakis(decyl)ammonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide

A248667-100G A248667-1G A248667-25G A248667-5G
76786-770EA 231 USD
76786-770 76786-772 76786-774 76786-776
Tetrakis(decyl)ammonium bromide 98%
Tetrakis(decyl)ammonium bromide

Precaución: for Research Use Only

Formula: C₄₀H₈₄BrN
MW: 659.0057 g/mol
MDL Number: MFCD00043166
CAS Number: 14937-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR