5,6-Dichloro-1H-indole 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloroindole

A248478-100G A248478-10G A248478-1G A248478-25G A248478-5G
76786-754EA 502.87 USD
76786-754 76786-756 76786-758 76786-760 76786-762
5,6-Dichloro-1H-indole 98%
5,6-Dichloro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MW: 186.0400 g/mol
MDL Number: MFCD01075036
CAS Number: 121859-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR