Ethyl-2-oxo-2-(pyridin-3-yl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxo-2-(pyridin-3-yl)acetate

A248290-1G A248290-250MG
76786-736EA 211.12 USD
76786-736 76786-738
Ethyl-2-oxo-2-(pyridin-3-yl)acetate 98%
Ethyl-2-oxo-2-(pyridin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.1727 g/mol
MDL Number: MFCD09842445
CAS Number: 73672-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR